Please login to your online account to display your discounted pricing

Ferric Ammonium Sulfate, Dodecahydrate, ACS Grade, 98.5% to 102.0%, LabChem™

$147.11 - $147.11

Chemical Identifiers

Chemical Name or Material Ferric Ammonium Sulfate
CAS 7783-83-7
CAS Max % 100
Name Note Dodecahydrate
Purity Grade ACS Grade
Linear Formula FeNH4S2O8·12H2O
Formula Weight 482.19
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC142601

 
lab chem
LC142601
500g Each for $147.11
Description & Specifications

Specifications

Formula Weight 482.19
Assay 98.5 to 102.0%
Identification Passes Test
Health Hazard 1 Warning
Purity Grade ACS Grade
Solubility Soluble in water
EINECS Number 233-382-4
CAS Max % 100
Linear Formula FeNH4S2O8·12H2O
CAS 7783-83-7
Specific Gravity 1710kg/m3
Chemical Name or Material Ferric Ammonium Sulfate
Name Note Dodecahydrate
Melting Point 39°C
Color Violet
Quantity 500g
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Packaging Poly Bottle
Physical Form Solid
Decomposition Information Nitrogen oxides; Gaseous Ammonia; Sulfur compounds
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Density 1,710kg/m3
Storage Information Room Temperature
Safety and Handling

Warning

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.