Please login to your online account to display your discounted pricing
Fernbach Flask Clamps for Sartorius™ Certomat™ Benchtop Shakers
In stainless steel, for a variety of flask sizes
$89.60 - $141.40
Specifications
For Use With Certomat™ MOII RM SII IS, BS-1, BS-T, CT Plus
Type Clamp
Material Stainless Steel
View More Specs
Products
Catalog Number Mfr. No. Holds Price Quantity    
14-559-161
sartorius stedim
BBI8854600
1800mL Fernbach Flask Each for $89.60
14-559-165
sartorius stedim
BBI8854640
2800mL Fernbach Flask Each for $141.40
Description & Specifications
Specifications
For Use With Certomat™ MOII RM SII IS, BS-1, BS-T, CT Plus
Type Clamp
Material Stainless Steel