Please login to your online account to display your discounted pricing
Thermo Scientific™ TurboFect Transfection Reagent
Get efficient in vitro delivery of plasmid DNA to cultured cells with low cytotoxicity.
$316.00 - $556.00
Specifications
For Use With (Application) DNA transfection.siRNA-DNA co-transfection.
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    
FERR0531
thermo scientific™
R0531
1mL Each for $316.00
FERR0541
thermo scientific™
R0541
120µL Each for $556.00
Description & Specifications
Specifications
For Use With (Application) DNA transfection.siRNA-DNA co-transfection.
TurboFect Transfection Reagent is recommended for efficient transfection of plasmid DNA. Distinct from lipid-based formulas, TurboFect is a sterile solution of a proprietary cationic polymer in water. The polymer forms positively-charged complexes with DNA that are both compact and stable. These complexes protect DNA from degradation and facilitate efficient plasmid delivery into eukaryotic cells.
Highlights
  • High transfection efficiency of plasmid DNA into a wide variety of cell types
    • Adherent and suspension cells
    • Differentiated and undifferentiated cells
  • Excellent transfection efficiency in the presence or absence of serum 
  • Ready-to-use reagent - no need to reconstitute, dilute, or manipulate
  • Simple protocol
Review our experimental considerations to improve your plasmid transfection results.
Review our experimental considerations to improve your plasmid transfection results.