Thermo Scientific™ SP6 RNA Polymerase
Catalyze the 5'→3' synthesis of RNA on either single-stranded or double-stranded DNA with Thermo Scientific™ SP6 RNA polymerase.
$77.00 - $173.00
Specifications
Description SP6 RNA Polymerase
For Use With (Application) Synthesis of unlabeled and labeled RNA that can be used: for hybridization (1), in vitro RNA translation (2); as aRNA (3), siRNA (4), substrate in RNase protection assays (5), template for genomic DNA sequencing.
Includes 1mL of 5X Transcription Buffer
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
FEREP0131 Thermo Scientific™
EP0131
2,000U Each for $77.00
FEREP0133 Thermo Scientific™
EP0133
1000U Each for $173.00
Specifications
Description
Bacteriophage SP6 RNA polymerase is a DNA-dependent RNA polymerase with strict specificity for its respective double-stranded promoters. It catalyzes the 5'→3' synthesis of RNA on either single-stranded DNA or double-stranded DNA downstream from its promoter and incorporates modified nucleotides.
Highlights
  • Incorporates modified nucleotides (e.g., aminoallyl-, biotin-, fluorescein-, digoxigenin-labeled nucleotides)
Description & Specifications
Specifications
SP6 RNA Polymerase
Synthesis of unlabeled and labeled RNA that can be used: for hybridization (1), in vitro RNA translation (2); as aRNA (3), siRNA (4), substrate in RNase protection assays (5), template for genomic DNA sequencing.
1mL of 5X Transcription Buffer