Please login to your online account to display your discounted pricing
Thermo Scientific™ PagI (BspHI)
The PagI (BspHI) restriction enzyme recognizes T^CATGA sites and cuts best at 37°C in O buffer (Isoschizomers: BspHI, CciI, RcaI).
$72.00 - $292.00
Specifications
Concentration 10U/µL
Product Name PagI (BspHI)
View More Specs
Products
Catalog Number Mfr. No. Units Price Quantity    
FERER1281
thermo scientific™
ER1281
400U Each for $72.00
FERER1282
thermo scientific™
ER1282
2,000U Each for $292.00
Description & Specifications
Specifications
Concentration 10U/µL
Product Name PagI (BspHI)
Lambda DNA digested with PagI (BspHI), 0.7% agarose, 3 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities