Thermo Scientific™ Taq DNA Polymerase, Native
Optimize amplification of DNA samples of low purity with this highly thermostable, native DNA polymerase for routine PCR.
$219.00 - $219.00
Specifications
Description Taq™
For Use With (Application) Routine PCR amplification of DNA fragments up to 5kb (1); Generation of PCR product for TA cloning;DNA labeling (2–4); DNA sequencing (5)
View More Specs
Disclaimers:
In certain countries use of this product is covered by patents. Purchase of product in these countries includes non-transferable, limited license for using only this amount of product for the purchaser's own internal research.
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
FEREP0282 Thermo Scientific™
EP0282
2 x 1mL of 25mM MgCl2, 2 x 1mL of 10X Taq Buffer with KCl, 2 x 1mL of 10X Taq Buffer with (NH4)2SO4 Each for $219.00
Specifications
Description
Native Taq DNA Polymerase is a highly thermostable DNA polymerase of the thermophilic bacterium Thermus aquaticus. The enzyme catalyzes 5'→3' synthesis of DNA, has no detectable 3'→5' exonuclease (proofreading) activity and possesses low 5'→3' exonuclease activity. In addition, the polymerase exhibits deoxynucleotidyl transferase activity, which frequently results in the addition of extra adenines at the 3'-end of PCR products. Native Taq DNA Polymerase is preferred for amplification of bacterial DNA sequences homologous to those found in E. coli.
Taq DNA Polymerase (native, with BSA) is supplied with BSA as a stabilizing agent. This version of Taq DNA Polymerase is often the best choice when amplifying DNA samples of lower purity, e.g. genomic DNA from mouse tail.
Highlights
  • Thermostable -half life is more than 40 min at 95°C.
  • Generates PCR products with 3'-dA overhangs.
  • Supplied with two buffers -10X Taq Buffer with KCl and 10X Taq Buffer with (NH4)2SO4. The latter allows for PCR at wide range of magnesium concentrations and decreases unspecific priming.
  • Incorporates modified nucleotides (e.g., biotin-, digoxigenin-, fluorescently-labeled nucleotides).
Description & Specifications
Specifications
Taq™
Routine PCR amplification of DNA fragments up to 5kb (1); Generation of PCR product for TA cloning;DNA labeling (2–4); DNA sequencing (5)