Please login to your online account to display your discounted pricing
Thermo Scientific™ Maxima Hot Start Taq DNA Polymerase
Enhance the specificity, sensitivity, and yield of DNA amplification with this hot start DNA polymerase.
Manufacturer: thermo scientific™  EP0602
 View more versions of this product
Catalog No. FEREP0602
  • / Each
Description & Specifications
Specifications
Description Maxima™ Hot Start™ Taq DNA Polymerase
For Use With (Application) Hot start PCR; High yield amplification of targets up to 3kb; RT-PCR.
Quantity 100U
Includes 2 x 1.25mL of 10X Hot Start PCR Buffer, 2 x 1.25mL of 25mM MgCl2
Maxima Hot Start Taq DNA Polymerase is a chemically modified recombinant Taq DNA polymerase. The enzyme is inactive at room temperature, avoiding extension of non-specifically annealed primers or primer dimers and providing higher specificity of DNA amplification. The functional activity of the enzyme is restored during a short 4 minute incubation at 95°C. The activated enzyme maintains the same functionality as Taq DNA polymerase. Maxima Hot Start Taq DNA Polymerase is also available in a master mix format.
In addition, the enzyme provides the convenience of reaction set-up at room temperature. Maxima Hot Start Taq DNA Polymerase is a recombinant Taq DNA polymerase which has been chemically modified by the addition of heat-labile blocking groups to its amino acid residues. The enzyme is inactive at room temperature, avoiding extension of non-specifically annealed primers or primer dimers and providing higher specificity of DNA amplification (see Fig. 1). The functional activity of the enzyme is restored during a short 4-minute incubation at 95°C. The activated enzyme maintains the same functionality as Taq DNA polymerase: catalyzes 5'=>3' synthesis of DNA, has no detectable 3'=>5' proofreading exonuclease activity, but possesses low 5'=>3' exonuclease activity. It exhibits deoxynucleotidyl transferase activity, which frequently results in the addition of extra adenines at the 3'-end of PCR products. Before activation, the two activities are not detectable.
  • High yield amplification of complex templates
  • Four minutes activation time
  • High PCR specificit—reduced effects of mispriming and primer-dimer formation
  • Enhanced PCR sensitivity
  • Convenient room temperature PCR set-up
  • Generates PCR products with 3'-dA overhangs
Recommended for:
Hot start PCR; High yield amplification of targets up to 3kb; RT-PCR.