Please login to your online account to display your discounted pricing

Thermo Scientific™ Maxima™ Probe 2X qPCR Master Mix with ROX Solution


Optimize qPCR with probe chemistry using standard cycling protocols with these ready-to-use qPCR master mixes.

Manufacturer: thermo scientific™  K0262

 View more versions of this product

 Disclaimers

A license to perform the patented 5' Nuclease Process for research is obtained by the purchase of (i) both Authorized 5' Nuclease Core Kit and Licensed Probe, (ii) a Licensed 5' Nuclease Kit, or (iii) license rights from Applied Biosystems. This product is an Authorized 5' Nuclease Core Kit. Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: 5,789,224, 5,618,711, 6,127,155, 5,677,152, 5,773,258, 5,407,800, 5,322,770, 5,310,652. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser's own internal research. Separate purchase of a Licensed Probe would convey rights under the applicable claims of US Patents Nos. 5,538,848, 5,723,591, 5,876,930, 6,030,787, 6,258,569, 5,804,375 (claims 1-12 only), and corresponding claims outside the United States. No right under any other patent claim and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser's activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained from the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA. Use of this product in a passive reference method is covered by the following U.S. Patent: 5,928,907 (claim numbers 12-24, 27-28) and corresponding patent claims outside the US. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product in a passive reference method for the purchaser's own internal research. No right under any other patent claim and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser's activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.

Use of UDG for PCR carryover decontamination is covered by patents in certain countries and may require a license.

Catalog No. FERK0262

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Real-time PCR and RT-qPCR using sequence specific probes on most real-time PCR machines.
Quantity 10 x 1.25mL
No. of Reactions 1000 reactions of 25 µl
Description Maxima™ Probe™ qPCR Master Mix (2X), ROX Solution provided

Maxima Probe qPCR Master Mixes are ready-to-use solutions optimized for quantitative real-time PCR. The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added.

Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG). The use of Maxima Probe qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates.

Maxima Probe qPCR Master Mixes are compatible with most real-time thermal cyclers.

The master mix includes Maxima Hot Start Taq DNA polymerase and dNTPs in an optimized PCR buffer. ROX Solution is provided in a separate vial for use with machines that require ROX as a passive reference dye. Maxima Hot Start Taq DNA polymerase in combination with the optimized buffer ensures PCR specificity and sensitivity.dUTP is included in the mix for optional carryover contamination control using uracil-DNA glycosylase (UDG). The use of Maxima Probe qPCR Master Mix in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral and cDNA templates.

  • Specificity—Maxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
  • Sensitivity—detects low copy number targets
  • ROX Solution provided in a separate tube
  • Wide linear range—accurate quantification across 9 orders of magnitude
  • Universal—can be used on most real-time PCR instruments
  • Reproducibility and convenience—ready-to-use 2X master mix

Recommended for:

Real-time PCR and RT-qPCR using sequence specific probes on most real-time PCR machines.