Thermo Scientific™ GeneRuler™ DNA Ladder Mix
Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels.
$163.00 - $654.00
Specifications
Quantity 5 × 50μg
View More Specs
Disclaimers:
This product or its use is covered by at least one claim of U.S. Patent Nos. 5,834,201, 6,680,378, and/or 7,132,520 owned by Invitrogen Corporation. The purchase of this product conveys to the buyer the non-transferable right to use the purchased amount of the product in internal research conducted by the buyer. The buyer cannot use this product or materials made by the employment of this product for Commercial Purposes. Commercial Purposes means any activity for consideration and may include, but is not limited to: (1) use of the product in manufacturing; (2) use of the product to provide a service, information, or data; (3) use of the product for therapeutic, diagnostic or prophylactic purposes; or (4) resale of the product, whether or not the product is resold for use in research. Further information on purchasing licenses under the above patents may be obtained by contacting the Business Development Department, Invitrogen Corporation, 5791 Van Allen Way, Carlsbad, CA 92008.
Products ${productFamilyLength}
Catalog Number Mfr. No. Kit Contents Price Quantity  
Catalog Number Mfr. No. Kit Contents Price Quantity  
FERSM0331 Thermo Scientific™
SM0331
6X DNA Loading Dye, 2 x 1mL Each for $163.00
FERSM0332 Thermo Scientific™
SM0332
6X DNA Loading Dye, 10 x 1mL Each for $654.00
Specifications
Description
GeneRuler DNA Ladder Mix is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels. The ladder is a mixture of chromatography-purified individual DNA fragments.
GeneRuler DNA Ladder Mix can be labeled radioactively with T4 Polynucleotide Kinase. The ladder is supplied with 6X DNA Loading Dye.
Highlights
  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Bright reference bands
  • Supplied with loading dye for sample DNA
Description & Specifications
Specifications
5 × 50μg