Thermo Scientific™ BseDI (BsaJI)
Cut at C^CNNGG sites with BseDI (BsaJI) restriction enzyme, which performs best at 55°C in Tango buffer (Isoschizomers: BsaJI, BssECI).
$65.10 - $65.10
Specifications
Concentration 10U/µL
Product Name BseDI (BsaJI)
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
FERER1081 Thermo Scientific™
ER1081
300U Each for $65.10
Specifications
Description
Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Description & Specifications
Specifications
10U/µL
BseDI (BsaJI)