Please login to your online account to display your discounted pricing
Thermo Scientific™ Biotin Chromogenic Detection Kit
Perform chromogenic detection of biotinylated nucleic acid probes with the ThermoScientific™ Biotin Chromogenic Detection Kit.
$140.00 - $275.00
Specifications
For Use With (Application) Southern, Northern, dot/slot blots, plaque or colony screening.
Includes Streptavidin-AP Conjugate;BCIP/NBT Solution;Blocking Reagent;10X Blocking/Washing Buffer;10X Detection Buffer;Detailed Protocol
View More Specs
Products
Catalog Number Mfr. No. No. of Reactions Price Quantity    
FERK0661
thermo scientific™
K0661
10 Each for $140.00
FERK0662
thermo scientific™
K0662
30 Each for $275.00
Description & Specifications
Specifications
For Use With (Application) Southern, Northern, dot/slot blots, plaque or colony screening.
Includes Streptavidin-AP Conjugate;BCIP/NBT Solution;Blocking Reagent;10X Blocking/Washing Buffer;10X Detection Buffer;Detailed Protocol
Biotin Chromogenic Detection Kit is a convenient tool for the chromogenic detection of biotinylated nucleic acid probes. The kit is optimized to reproducibly provide high sensitivity with a low background in applications, such as Southern, Northern, dot and slot blotting, and screening of viral plaques and bacterial colonies. Biotinylation of DNA/RNA probes is widely used as a safe and convenient alternative to radioactive labeling. Biotin can be incorporated into nucleic acids using various enzymatic or non-enzymatic methods, including the Biotin DecaLabel DNA Labeling Kit. 
Highlights
Biotinylated probes are detected with streptavidin coupled to alkaline phosphatase (AP). Streptavidin-AP conjugates bind specifically and irreversibly to the biotin-labeled probes. The probes are then visualized using a chromogenic substrate for alkaline phosphatase BCIP/NBT, which produces a blue-purple precipitate. Visualization does not require X-ray film or other specific equipment.
Highlights
  • High sensitivity of detection - 0.03 pg of homologous DNA in dot blot hybridization with biotin-labeled probe
  • Low background due to optimized washing and blocking procedures
  • Non-radioactive detection visualization does not require X-ray film or other specific equipment
  • Fast and convenient - ready-to-use components