Thermo Scientific™ Aminoallyl-dUTP
Perform enzymatic indirect non-radioactive labeling of DNA with the modified nucleotide Aminoallyl-dUTP (5-[3-aminoallyl]-2'-deoxyuridine-5'-triphosphate).
$110.25 - $196.00
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
FERR0091 Thermo Scientific™
R0091
1µmol (0.1mL of 10mM solution) Each for $110.25
FERR1101 Thermo Scientific™
R1101
2.5µmol Each for $196.00
Specifications
Description
Aminoallyl-dUTP (5-[3-aminoallyl]-2'-deoxyuridine-5'-triphosphate) is supplied as 10 mM and 50 mM aqueous solutions titrated to pH 7.0 with NaOH.
Aminoallyl-dUTP can be enzymatically incorporated into DNA with Reverse Transcriptases, Taq DNA Polymerase, phi29 DNA Polymerase, Klenow Fragment, Klenow Fragment, exo- and DNA Polymerase I. The resulting amine-containing DNA can be subsequently labeled with any amine-reactive fluorescent dye, biotin or hapten.
Aminoallyl-dUTP can be enzymatically incorporated into DNA with Reverse Transcriptases, Taq DNA Polymerase, phi29 DNA Polymerase, Klenow Fragment, Klenow Fragment, exo- and DNA Polymerase I. The resulting amine-containing DNA can be subsequently labeled with any amine-reactive fluorescent dye, biotin or hapten.
Recommended For:
Enzymatic indirect non-radioactive labeling of DNA during cDNA synthesis, PCR, nick-translation, random primed labeling or primer extension.