Please login to your online account to display your discounted pricing

Thermo Scientific™ 10X Taq™

Find the right buffer to meet your specific PCR needs in these 10X Taq DNA Polymerase buffers with different compositions.

$21.05 - $21.05

Specifications

Components 100mM Tris-HCl (pH 8.8 at 25°C); 500mM KCl
Quantity 4 x 1.25mL
Storage Requirements -20°C
View More Specs
Products
Catalog Number Mfr. No. Product Name Price Quantity    

FERB16

 
thermo scientific™
B16*
10X Taq Buffer with KCl and 15mM MgCl2 Each for $21.05

FERB33

 
thermo scientific™
B33
10X Taq Buffer with (NH4)2SO4 Each for $21.05

FERB34

 
thermo scientific™
B34
10X Taq Buffer with (NH4)2SO4 and 20 mM MgCl2 Each for $21.05

FERB38

 
thermo scientific™
B38
10X Taq Buffer with KCl Each for $21.05

FERB55

 
thermo scientific™
B55
10X Taq Buffer without Detergent Each for $21.05
Description & Specifications

Specifications

Components 100mM Tris-HCl (pH 8.8 at 25°C); 500mM KCl
Quantity 4 x 1.25mL
Storage Requirements -20°C

10X Taq Buffers are ready-to-use buffers for PCR using Taq DNA Polymerases (both recombinant and native). Available in different compositions and also without detergents.

Quality Control

  • Tested for the absence of endo-, exodeoxyribonucleases, ribonucleases and functionally tested in amplification of 956 bp DNA fragment of a single-copy gene of human genomic DNA

Composition

  • 100mM Tris-HCl (pH 8.8 at 25°C)
  • 500mM KCl