missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Taq Buffer (10X), with KCl
Thermo Scientific 10X Taq Buffers are ready-to-use buffers for PCR using Taq DNA Polymerases (both recombinant and native).
Supplier: Thermo Scientific™ B38
Description
Thermo Scientific™ 10X Taq Buffer without Detergent is used in PCR with Fermentas Taq DNA Polymerases (both recombinant and native), and this buffer does not contain detergents which can interfere with certain applications, e.g. microarray experiments. Additional Information and Online Ordering
- Thermo Scientific 10X Taq Buffers are ready-to-use buffers for PCR using Taq DNA Polymerases (both recombinant and native)
- Available in different compositions and also without detergents
- Store at -20°C
10X Taq Buffer with KCl (B38) includes:
- 100 mM Tris-HCl (pH 8.8 at 25°C)
- 500 mM KCl
- Detergent
Specifications
Contains: • 4 x 1.25 mL 10X Taq Buffer with KCl Store at -20°C. |
|
Taq DNA Polymerase | |
10X Taq Buffer with KCl | |
10X |
Liquid | |
Thermo Scientific™ | |
PCR |
For Research Use Only. Not for use in diagnostic procedures.