Please login to your online account to display your discounted pricing

Thermo Scientific™ 10X Taq™


Find the right buffer to meet your specific PCR needs in these 10X Taq DNA Polymerase buffers with different compositions.

Manufacturer: thermo scientific™  B34

 View more versions of this product

Catalog No. FERB34

  • / Each
 

Description & Specifications

Specifications

Product Name 10X Taq Buffer with (NH4)2SO4 and 20 mM MgCl2
Quantity 4 x 1.25mL
Storage Requirements -20°C
Components 100mM Tris-HCl (pH 8.8 at 25°C); 500mM KCl
Product Name 10X Taq Buffer with (NH4)2SO4 and 20 mM MgCl2
Quantity 4 x 1.25mL
Item Description 4 x 1.25 mL; W/(NH4)2SO4 and 20 mM MgCl2

10X Taq Buffers are ready-to-use buffers for PCR using Taq DNA Polymerases (both recombinant and native). Available in different compositions and also without detergents.

Quality Control

  • Tested for the absence of endo-, exodeoxyribonucleases, ribonucleases and functionally tested in amplification of 956 bp DNA fragment of a single-copy gene of human genomic DNA

Composition

  • 100mM Tris-HCl (pH 8.8 at 25°C)
  • 500mM KCl