Please login to your online account to display your discounted pricing

FERb 033, Tocris™

Potent and selective ERβ agonist

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material FERb 033
Assay Percent Range >98%
Molecular Formula C13H9ClFNO3
Formula Weight 281.67
Synonym 2-Chloro-3'-fluoro-3, 4'-dihydroxy-[1, 1-biphenyl]-4-carboxaldehyde oxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

352310

 
r & d systems
RDS352310
10mg Each for $214.12

352350

 
r & d systems
RDS352350
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 281.67
Quantity 10mg
Recommended Storage Store at +4°C
Molecular Formula C13H9ClFNO3
CAS 1111084-78-6
Chemical Name or Material FERb 033
Synonym 2-Chloro-3'-fluoro-3, 4'-dihydroxy-[1, 1-biphenyl]-4-carboxaldehyde oxime
Assay Percent Range >98%
Safety and Handling

Store at +4°C