Invitrogen™ FER Recombinant Human Protein DFS Item
FER is a member of the FPS/FES family of non-receptor tyrosine kinases.
Manufacturer:  Invitrogen™ LSPV4076
 View more versions of this product
Catalog No. PV4076
Description
Description
FER regulates cell-cell adhesion and mediates signaling from the cell surface to the cytoskeleton via growth factor receptors.
Specifications
Specifications
NP_005237
Frozen
60.3kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Catalytic
Kinase Assay
FER Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton™ X-100, 2mM DTT and 50% glycerol
1mg
RUO - research use only
FER, TYK3
Baculovirus
Human
His Tag
Documents