Please login to your online account to display your discounted pricing

Alfa Aesar™ Fenvalerate, 99%

Type II pyrethrin and a potent inhibitor of calcineurin (protein phosphatase 2B)

Chemical Identifiers

Chemical Name or Material Fenvalerate
CAS 51630-58-1
Molecular Formula C25H22ClNO3
Formula Weight 419.91
MDL Number MFCD00055324
UN Number UN2811
Beilstein 2025982
Merck Index 14,4010
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ63562FPL

  SDS
alfa aesar™
J63562FPL
20mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ63562MC

  SDS
alfa aesar™
J63562MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Fenvalerate
Color White
Melting Point 54° to 59°C
Quantity 20mg
CAS 51630-58-1
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage −30° to −10°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number CY1576350
Physical Form Waxy Solid
Assay 99%
Molecular Formula C25H22ClNO3
Formula Weight 419.91
MDL Number MFCD00055324
UN Number UN2811
Beilstein 2025982
Merck Index 14,4010

Applications
Fenvalerate is used to control insects from food crops, animal feed and cotton products. Control of a wide range of pests, including those resistant to organochlorine, organophosphorus, and carbamate.

Solubility
Soluble in 100% ethanol (>25 mg/ml), water (0.001 g/L) at 20°C, and DMSO.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, acids, heat.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

−30° to −10°C