Please login to your online account to display your discounted pricing

Fenretinide, Tocris

Synthetic retinoid; Potent anticancer agent

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material Fenretinide
Assay Percent Range >98%
Molecular Formula C26H33NO2
Formula Weight 391.55
Synonym N-(4-Hydroxyphenyl)retinamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

139610

 
r & d systems
RDS139610
10mg Each for $177.37

139650

 
r & d systems
RDS139650
50mg Each for $746.48
Description & Specifications

Specifications

Formula Weight 391.55
Quantity 10mg
Molecular Formula C26H33NO2
Assay Percent Range >98%
Synonym N-(4-Hydroxyphenyl)retinamide
Chemical Name or Material Fenretinide
CAS 65646-68-6
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C