Please login to your online account to display your discounted pricing

Alfa Aesar™ (+)-Fenchone, 98%

Chemical Identifiers

Chemical Name or Material (+)-Fenchone
CAS 4695-62-9
Molecular Formula C10H16O
Formula Weight 152.24
MDL Number MFCD00070689
UN Number UN1224
Beilstein 2206555
Merck Index 14,3968
Synonym (1S)-(+)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one; (+)-1,3,3-Trimethylnorcamphor
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1593814

  SDS
alfa aesar™
L1593814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1593822

  SDS
alfa aesar™
L1593822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 152.24
Chemical Name or Material (+)-Fenchone
Molecular Formula C10H16O
CAS 4695-62-9
Assay 98%
MDL Number MFCD00070689
Beilstein 2206555
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
UN Number UN1224
Merck Index 14,3968
Synonym (1S)-(+)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one; (+)-1,3,3-Trimethylnorcamphor
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
RTECS Number RB7875200
Flash Point 52°C (125°F)
Optical Rotation +60° (Neat)
Boiling Point 192° to 194°C
Melting Point 5° to 7°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.