Please login to your online account to display your discounted pricing

Felodipine, Tocris

Ca2+ channel blocker (L-type)

$133.98 - $567.83

Chemical Identifiers

Chemical Name or Material Felodipine
Assay Percent Range >98%
Molecular Formula C18H19Cl2NO4
Formula Weight 384.25
Synonym 4-(2,3-Dichlorophenyl)-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic acid ethyl methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

296010

 
r & d systems
RDS296010
10mg Each for $133.98

296050

 
r & d systems
RDS296050
50mg Each for $567.83
Description & Specifications

Specifications

Formula Weight 384.25
Quantity 10mg
Chemical Name or Material Felodipine
Assay Percent Range >98%
Molecular Formula C18H19Cl2NO4
CAS 72509-76-3
Synonym 4-(2,3-Dichlorophenyl)-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic acid ethyl methyl ester
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C