Please login to your online account to display your discounted pricing

Felbamate, Tocris


NMDA antagonist, acts glycine site

Quantity 10mg

Manufacturer: r & d systems  RDS086910

 View more versions of this product

Catalog No. 086910

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 238.24
Molecular Formula C11H14N2O4
Assay Percent Range >99%
CAS 25451-15-4
Synonym 2-Phenyl-1, 3-propanedioldicarbamate
Recommended Storage Store at Room Temperature
Chemical Name or Material Felbamate

Specifications

Chemical Name or Material Felbamate
Assay Percent Range >99%
Molecular Formula C11H14N2O4
Formula Weight 238.24
Synonym 2-Phenyl-1, 3-propanedioldicarbamate
Accessories

Store at Room Temperature