Please login to your online account to display your discounted pricing

Felbamate, Tocris

NMDA antagonist, acts glycine site

$139.08 - $585.69

Chemical Identifiers

Chemical Name or Material Felbamate
Assay Percent Range >99%
Molecular Formula C11H14N2O4
Formula Weight 238.24
Synonym 2-Phenyl-1, 3-propanedioldicarbamate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

086910

 
r & d systems
RDS086910
10mg Each for $139.08

086950

 
r & d systems
RDS086950
50mg Each for $585.69
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 238.24
Assay Percent Range >99%
Molecular Formula C11H14N2O4
CAS 25451-15-4
Synonym 2-Phenyl-1, 3-propanedioldicarbamate
Chemical Name or Material Felbamate
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature