missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fehling's Solution 'B', Alkaline No. 2, Certified, LabChem™

Manufacturer:  LabChem LC142106

 View more versions of this product

Catalog No. LC142106



Specifications

Specifications

Fehling's Solution 'B', Alkaline No. 2
60.9,34.6,4.5
KNaC4H4O6·4H2O
Passes Test
Liquid
Cubitainer™
Soluble in water
Colorless
Alkaline No. 2
6381-59-5,1310-73-2
UN1824
282.22
Certified
Passes Test
Sodium oxide; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
10L
Documents
Promotions

Promotions