Please login to your online account to display your discounted pricing

Febuxostat, Tocris™

Xanthine oxidase inhibitor

$221.01 - $895.51

Chemical Identifiers

Chemical Name or Material Febuxostat
Assay Percent Range >99%
Molecular Formula C16H16N2O3S
Formula Weight 316.37
Synonym 2-[3-Cyano-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

442710

 
r & d systems
RDS442710
10mg Each for $221.01

442750

 
r & d systems
RDS442750
50mg Each for $895.51
Description & Specifications

Specifications

Formula Weight 316.37
Quantity 10mg
CAS 144060-53-7
Synonym 2-[3-Cyano-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic acid
Molecular Formula C16H16N2O3S
Assay Percent Range >99%
Chemical Name or Material Febuxostat
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature