Please login to your online account to display your discounted pricing

FDC Mouse anti-Human, Biotin, Clone: CNA.42, eBioscience™

Mouse Monoclonal Antibody

$140.65 - $353.80

Specifications

Antigen FDC
Clone CNA.42
Host Species Murine
Gene Alias Follicular dendritic cell
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122623

 
life technologies
LS13996880
25μg Each for $140.65

501122624

 
life technologies
LS13996882
100μg Each for $353.80
Description & Specifications

Specifications

Antigen FDC
Clone CNA.42
Host Species Murine
Gene Alias Follicular dendritic cell
Species Reactivity Human
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunohistochemistry (Frozen)
Applications Western Blot
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody CNA.42 recognizes a unique carbohydrate epitope found on follicular dendritic cells (FDCs) from a variety of species. FDCs are large cells in germinal centers (GCs) of primary and lymphoid tissues. They present non-processed antigen and switch off apoptosis of B cells through a selection process resulting in selection of memory B cells. FDCs express many B cell markers such as CD21 (long isoform), CD23 and CD35. A role for FDCs as been shown in various infectious diseases such as AIDS and transmissible spongiform encephalitis. Staining with this CNA.42 monoclonal antibody is mainly restricted to the plasma membrane but in limited cases may also be cytoplasmic (as seen in mast cells).

This monoclonal has been shown to be a useful reagent for both flow cytometry and IHC analysis.