Please login to your online account to display your discounted pricing

FCCP, Tocris


Oxidative phosphorylation uncoupler

Quantity 10mg

Manufacturer: r & d systems  RDS045310

 View more versions of this product

Catalog No. 045310

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 254.17
Chemical Name or Material FCCP
Assay Percent Range >99%
Molecular Formula C10H5F3N4O
CAS 370-86-5
Synonym Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material FCCP
Assay Percent Range >99%
Molecular Formula C10H5F3N4O
Formula Weight 254.17
Synonym Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone
Accessories

Store at Room Temperature