Fc epsilon Receptor 1 alpha (FceRI) Mouse anti-Human, PE-eFluor 610, Clone: AER-37 (CRA1), eBioscience™
Mouse Monoclonal Antibody
Manufacturer:  Life Technologies LS61589942
 View more versions of this product
Catalog No. 501124849
Description
Description
The AER-37 monoclonal antibody reacts with the Fc epsilon RI alpha subunit, an IgE-binding subunit lacking signal-transducing ability. Fc epsilon RI apha is expressed on mast and basophil cells and is upregulated by the presence of IgE. Fc epsilon RI alpha forms a tetrameric complex with one beta and two gamma subunits. The beta and gamma subunits possess immunoreceptor tyrosine-based activation motifs (ITAM). The Fc epsilon RI complex plays an important role in triggering IgE-mediated allergic reactions.
Specifications
Specifications
Fc epsilon Receptor 1 alpha (FceRI)
AER-37 (CRA1)
PE-eFluor 610
Conjugated
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Monoclonal
Flow Cytometry
5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
FceRI alpha, high affinity IgE receptor
IgG2b, kappa
RUO
Primary
Human
Documents
For Research Use Only.