Please login to your online account to display your discounted pricing

Fc epsilon Receptor 1 alpha (FceRI) Mouse anti-Human, PE-Cy7, Clone: AER-37 (CRA1), eBioscience™

Mouse Monoclonal Antibody

$205.90 - $516.20

Specifications

Antigen Fc epsilon Receptor 1 alpha (FceRI)
Clone AER-37 (CRA1)
Host Species Mouse
Gene Alias FceRI alpha, high affinity IgE receptor
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123409

 
life technologies
LS25589941
25 tests Each for $205.90

501123410

 
life technologies
LS25589942
100 tests Each for $516.20
Description & Specifications

Specifications

Antigen Fc epsilon Receptor 1 alpha (FceRI)
Clone AER-37 (CRA1)
Host Species Mouse
Gene Alias FceRI alpha, high affinity IgE receptor
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The AER-37 monoclonal antibody reacts with the Fc epsilon RI alpha subunit, an IgE-binding subunit lacking signal-transducing ability. Fc epsilon RI apha is expressed on mast and basophil cells and is upregulated by the presence of IgE. Fc epsilon RI alpha forms a tetrameric complex with one beta and two gamma subunits. The beta and gamma subunits possess immunoreceptor tyrosine-based activation motifs (ITAM). The Fc epsilon RI complex plays an important role in triggering IgE-mediated allergic reactions.