missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fc epsilon Receptor 1 alpha (FceRI) Mouse anti-Human, eFluor 450, Clone: AER-37 (CRA1), eBioscience™

Mouse Monoclonal Antibody

$161.04 - $402.60

Specifications

Antigen Fc epsilon Receptor 1 alpha (FceRI)
Clone AER-37 (CRA1)
Host Species Murine
Gene Alias FceRI alpha, high affinity IgE receptor
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124185 Life Technologies
LS48589942
100 Tests Each for $402.60
501124184 Life Technologies
LS48589941
25 Tests Each for $161.04
Specifications

Description

The AER-37 monoclonal antibody reacts with the Fc epsilon RI alpha subunit, an IgE-binding subunit lacking signal-transducing ability. Fc epsilon RI apha is expressed on mast and basophil cells and is upregulated by the presence of IgE. Fc epsilon RI alpha forms a tetrameric complex with one beta and two gamma subunits. The beta and gamma subunits possess immunoreceptor tyrosine-based activation motifs (ITAM). The Fc epsilon RI complex plays an important role in triggering IgE-mediated allergic reactions.
Description & Specifications

Specifications

Fc epsilon Receptor 1 alpha (FceRI)
Murine
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test
AER-37 (CRA1)
FceRI alpha, high affinity IgE receptor
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer