missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fc epsilon Receptor 1 alpha (FceRI) Mouse anti-Human, Clone: AER-37 (CRA1), eBioscience™

Mouse Monoclonal Antibody

$158.05 - $361.05

Specifications

Antigen Fc epsilon Receptor 1 alpha (FceRI)
Clone AER-37 (CRA1)
Host Species Murine
Gene Alias FceRI-a, FceRI-alpha, FceRI alpha, high affinity IgE receptor
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013309 Life Technologies
LS14589980
25μg Each for $158.05
501129417 Life Technologies
LS14589982
100μg Each for $361.05
Specifications

Description

The AER-37 monoclonal antibody reacts with the FcεRIα subunit, an IgE-binding subunit lacking signal-transducing ability. FcεRIα is expressed on mast and basophil cells and is upregulated by the presence of IgE. FcεRIα forms a tetrameric complex with one β and two γ subunits. The β and γ subunits possess immunoreceptor tyrosine-based activation motifs (ITAM). The FcεRI complex plays an important role in triggering IgE-mediated allergic reactions.
Description & Specifications

Specifications

Fc epsilon Receptor 1 alpha (FceRI)
Murine
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
AER-37 (CRA1)
FceRI-a, FceRI-alpha, FceRI alpha, high affinity IgE receptor
Flow Cytometry
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL