Please login to your online account to display your discounted pricing

Fc epsilon Receptor 1 alpha (FceRI) Armenian Hamster anti-Mouse, PE-eFluor 610, Clone: MAR-1, eBioscience™

Armenian Hamster Monoclonal Antibody

$172.55 - $404.55

Specifications

Antigen Fc epsilon Receptor 1 alpha (FceRI)
Clone 1-Mar
Host Species Armenian Hamster
Gene Alias high affinity IgE receptor
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124846

 
life technologies
LS61589880
25μg Each for $172.55

501124847

 
life technologies
LS61589882
100μg Each for $404.55
Description & Specifications

Specifications

Antigen Fc epsilon Receptor 1 alpha (FceRI)
Clone 1-Mar
Host Species Armenian Hamster
Gene Alias high affinity IgE receptor
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-eFluor 610
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The MAR-1 monoclonal antibody reacts with the Fc epsilon Receptor I alpha subunit, an IgE-binding subunit lacking signal-transducing ability. Fc epsilon RI alpha is expressed on mast and basophil cells and is up-regulated by the presence of IgE. Fc epsilon RI alpha forms a tetrameric complex with one beta and two gamma subunits. The beta and gamma subunits possess immunoreceptor tyrosine-based activation motifs (ITAM). The Fc epsilon RI complex plays an important role in triggering IgE-mediated allergic reactions.