missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fc epsilon Receptor 1 alpha (FceRI) Armenian Hamster anti-Mouse, APC-eFluor 780, Clone: MAR-1, eBioscience™

Armenian Hamster Monoclonal Antibody

$183.48 - $443.52

Specifications

Antigen Fc epsilon Receptor 1 alpha (FceRI)
Clone 1-Mar
Host Species Armenian Hamster
Gene Alias high affinity IgE receptor
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124063 Life Technologies
LS47589880
25μg Each for $183.48
501124064 Life Technologies
LS47589882
100μg Each for $443.52
Specifications

Description

The MAR-1 monoclonal antibody reacts with the Fc epsilon Receptor I alpha subunit, an IgE-binding subunit lacking signal-transducing ability. Fc epsilon RI alpha is expressed on mast and basophil cells and is up-regulated by the presence of IgE. Fc epsilon RI alpha forms a tetrameric complex with one beta and two gamma subunits. The beta and gamma subunits possess immunoreceptor tyrosine-based activation motifs (ITAM). The Fc epsilon RI complex plays an important role in triggering IgE-mediated allergic reactions.
Description & Specifications

Specifications

Fc epsilon Receptor 1 alpha (FceRI)
Armenian Hamster
Flow Cytometry
APC-eFluor 780
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL
1-Mar
high affinity IgE receptor
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer