missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fat red 7B, MP Biomedicals™

Manufacturer:  Mp Biomedicals 15803610

 View more versions of this product

Catalog No. ICN15803610


 

Specifications Description & Specifications

Specifications

Powder
Maroon
Molecular Biology
6368-72-5
Solvent Red 19
379.5
10g
Fat red 7B
C24H21N5

Chemical Identifiers

6368-72-5
C24H21N5
Solvent Red 19
Powder
379.5
Fat red 7B, Molecular Biology

Safety and Handling

EINECSNumber : 228-862-5

Recommended Storage : Store at Room Temperature (15° to 30°C)