Please login to your online account to display your discounted pricing

Fat red 7B, MP Biomedicals™

$67.68 - $67.68

Chemical Identifiers

Chemical Name or Material Fat red 7B
Name Note Molecular Biology
CAS 6368-72-5
Physical Form Powder
Molecular Formula C24H21N5
Formula Weight 379.5
Synonym Solvent Red 19
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15803610

 
mp biomedicals
15803610
10g Each for $67.68
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 379.5
Color Maroon
Quantity 10g
EINECS Number 228-862-5
Recommended Storage Store at Room Temperature (15° to 30°C)
Chemical Name or Material Fat red 7B
Name Note Molecular Biology
Molecular Formula C24H21N5
CAS 6368-72-5
Synonym Solvent Red 19
Safety and Handling

Store at Room Temperature (15° to 30°C)