missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Fasudil monohydrochloride, 99+%

$385.53 - $866.68

Chemical Identifiers

CAS 105628-07-7
Molecular Formula C14H18ClN3O2S
Molecular Weight (g/mol) 327.827
MDL Number MFCD00943198
InChI Key LFVPBERIVUNMGV-UHFFFAOYSA-N
Synonym fasudil hydrochloride, fasudil hcl, eril, fasudil hydrochloride, at 877 hydrochloride, fasudil monohydrochloride, ha 1077 hydrochloride, unii-sq04n8s7br, 5-1,4-diazepan-1-yl sulfonyl isoquinoline hydrochloride, sq04n8s7br
PubChem CID 163751
IUPAC Name 5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;hydrochloride
SMILES C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60751MD View Documents Alfa Aesar™
J60751MD
250mg Each for $385.53
 
AAJ60751ME View Documents Alfa Aesar™
J60751ME
500mg Each for $576.76
 
AAJ6075103 View Documents Alfa Aesar™
J6075103
1g Each for $866.68
 
Description

Description

Solubility
Soluble in water and dimethyl sulfoxide. Insoluble in ethanol.

Notes
Incompatible with acids, bases and oxidizing agents.
Specifications

Chemical Identifiers

105628-07-7
327.827
LFVPBERIVUNMGV-UHFFFAOYSA-N
163751
C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl
C14H18ClN3O2S
MFCD00943198
fasudil hydrochloride, fasudil hcl, eril, fasudil hydrochloride, at 877 hydrochloride, fasudil monohydrochloride, ha 1077 hydrochloride, unii-sq04n8s7br, 5-1,4-diazepan-1-yl sulfonyl isoquinoline hydrochloride, sq04n8s7br
5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;hydrochloride

Specifications

White
105628-07-7
MFCD00943198
fasudil hydrochloride, fasudil hcl, eril, fasudil hydrochloride, at 877 hydrochloride, fasudil monohydrochloride, ha 1077 hydrochloride, unii-sq04n8s7br, 5-1,4-diazepan-1-yl sulfonyl isoquinoline hydrochloride, sq04n8s7br
C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl
327.827
327.83
≥99%
250mg
C14H18ClN3O2S
14,3942
LFVPBERIVUNMGV-UHFFFAOYSA-N
5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;hydrochloride
163751
Powder
Fasudil monohydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : HM4031166

Recommended Storage : −30° to −10°C