missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Fasudil dihydrochloride, 99+%

$283.76 - $960.74

Chemical Identifiers

CAS 203911-27-7
Molecular Formula C14H19Cl2N3O2S
Molecular Weight (g/mol) 364.285
MDL Number MFCD00153805
InChI Key NOXXIYDYFSNHDF-UHFFFAOYSA-N
Synonym fasudil dihydrochloride, ha-1077 dihydrochloride, ha 1077, dihydrochloride, 5-1,4-diazepan-1-yl sulfonyl isoquinoline dihydrochloride, 5-isoquinolinesulfonyl homopiperazine, 2hcl, 5-isoquinolinesulfonyl homopiperazine,2hcl, 1-5-isoquinolinylsulfonyl homopiperazine dihydrochloride, 1-5-isoquinoline-sulfonyl-homopiperazine, fasudil and ha 1077, dihydrochloride, ha-1077 dihydrochloride, solid hplc
PubChem CID 16219471
IUPAC Name 5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;dihydrochloride
SMILES C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60594MD View Documents Alfa Aesar™
J60594MD
250mg Each for $283.76
 
AAJ60594ME View Documents Alfa Aesar™
J60594ME
500mg Each for $496.57
 
AAJ6059403 View Documents Alfa Aesar™
J6059403
1g Each for $960.74
 
Description

Description

Applications
Fasudil dihydrochloride is used in the inhibition of contractile responses to potassium chloride, phenylephnne (PHE), prostaglandin and hepatic stellate cells spreading. It is also used in the formation of stress fibers and in medicine.

Solubility
Soluble in water and dimethylsulfoxide.

Notes
Incompatible with strong oxidizing agents. Sore in a cool place.
Specifications

Chemical Identifiers

203911-27-7
364.285
NOXXIYDYFSNHDF-UHFFFAOYSA-N
16219471
C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl.Cl
C14H19Cl2N3O2S
MFCD00153805
fasudil dihydrochloride, ha-1077 dihydrochloride, ha 1077, dihydrochloride, 5-1,4-diazepan-1-yl sulfonyl isoquinoline dihydrochloride, 5-isoquinolinesulfonyl homopiperazine, 2hcl, 5-isoquinolinesulfonyl homopiperazine,2hcl, 1-5-isoquinolinylsulfonyl homopiperazine dihydrochloride, 1-5-isoquinoline-sulfonyl-homopiperazine, fasudil and ha 1077, dihydrochloride, ha-1077 dihydrochloride, solid hplc
5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;dihydrochloride

Specifications

White
203911-27-7
MFCD00153805
fasudil dihydrochloride, ha-1077 dihydrochloride, ha 1077, dihydrochloride, 5-1,4-diazepan-1-yl sulfonyl isoquinoline dihydrochloride, 5-isoquinolinesulfonyl homopiperazine, 2hcl, 5-isoquinolinesulfonyl homopiperazine,2hcl, 1-5-isoquinolinylsulfonyl homopiperazine dihydrochloride, 1-5-isoquinoline-sulfonyl-homopiperazine, fasudil and ha 1077, dihydrochloride, ha-1077 dihydrochloride, solid hplc
C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl.Cl
364.285
364.29
≥99%
250mg
C14H19Cl2N3O2S
14,3942
NOXXIYDYFSNHDF-UHFFFAOYSA-N
5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;dihydrochloride
16219471
Powder
Fasudil dihydrochloride
Safety and Handling

Safety and Handling

RTECSNumber : HM4031166

Recommended Storage : −30° to −10°C