Learn More
Thermo Scientific™ FastDigest™ NcoI Promotion
The FastDigest NcoI restriction enzyme recognizes C^CATGG sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: Bsp19I).
$19.32 - $221.01
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
FERFD0573PM |
Thermo Scientific™
FD0573PM |
20μL |
Each for $19.32
|
|
FERFD0574PM |
Thermo Scientific™
PMFD0574 |
100μL |
Each for $78.94
|
|
FERFD0575PM |
Thermo Scientific™
PMFD0575 |
300μL |
Each for $221.01
|
|
Description
Lambda DNA digested with NcoI, 0.7% agarose, 4 cleavage sites
FastDigest enzymes are an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest and FastDigest Green reaction buffers.
The universal buffer allows rapid single-, double- or multiple DNA digestion within 5-15 minutes eliminating any need for buffer change or subsequent DNA clean-up steps. DNA modifying enzymes, such as Klenow Fragment, T4 DNA Ligase, alkaline phosphatases and T4 DNA Polymerase all have 100% activity in FastDigest Buffer. Therefore, enzymes for downstream applications can be directly added to the FastDigest reaction mix.
For additional convenience FastDigest Green Buffer includes a density reagent and two tracking dyes for direct loading of digestion reaction products on gels.
Short incubation times and optimal composition of the universal FastDigest Buffer eliminate star activity effects.
Highlights
- 100% activity of all FastDigest enzymes in the universal buffer
- 100% buffer compatibility with downstream applications
- Complete digestion in 5-15 minutes
- Direct loading on gels
- No star activity
- 176 FastDigest enzymes available
Specifications
FastDigest™ NcoI |