missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fast Green, HARLECO™, MilliporeSigma™

Manufacturer:  Emd Millipore Corporation 21012

Catalog No. M21012



Description

Description

Specifications

Specifications

Fast Green
2353-45-9
C37H34N2O1S3Na2
Light Green
[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
808.84
88.86
Certified Biological Stain
Certistain™, For Microscopy
C37H34N2Na2O10S3
MFCD00013053
XJBPDZVCYTYRIN-UHFFFAOYSA-L
disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium
16887
Powder
25g

Chemical Identifiers

2353-45-9
808.84
XJBPDZVCYTYRIN-UHFFFAOYSA-L
16887
[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
C37H34N2Na2O10S3
MFCD00013053
Light Green
disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium
Safety and Handling

Safety and Handling

Recommended Storage : Store at +5°C to +3°C

SDS
Documents