missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fast Green FCF, TCI America™

Manufacturer:  TCI America F009910G

 View more versions of this product

Catalog No. F009910G



Specifications

Specifications

Fast Green FCF
C37H34N2Na2O10S3
RZSYLLSAWYUBPE-UHFFFAOYSA-L
disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-5-hydroxybenzenesulfonate
16887
Crystal-Powder at 20°C
Undesignated
2353-45-9
MFCD00013053
CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=C(C=C(C=C5)O)S(=O)(=O)[O-].[Na+].[Na+]
808.843
808.84
≥85.0% (HPLC,E)
10g

Chemical Identifiers

2353-45-9
808.843
RZSYLLSAWYUBPE-UHFFFAOYSA-L
disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-5-hydroxybenzenesulfonate
C37H34N2Na2O10S3
MFCD00013053
16887
CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=C(C=C(C=C5)O)S(=O)(=O)[O-].[Na+].[Na+]
Safety and Handling

Safety and Handling

EINECSNumber : (5)-5228

RTECSNumber : BQ4425000

TSCA : Yes

Documents