missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Fast Garnet GBC base

$28.38 - $98.24

Chemical Identifiers

CAS 97-56-3
Molecular Formula C14H15N3
Molecular Weight (g/mol) 225.295
MDL Number MFCD00007733
InChI Key PFRYFZZSECNQOL-UHFFFAOYSA-N
Synonym C.I. 11160; Solvent Yellow 3
PubChem CID 7340
IUPAC Name 2-methyl-4-[(2-methylphenyl)diazenyl]aniline
SMILES CC1=CC=CC=C1N=NC2=CC(=C(C=C2)N)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6057422 View Documents Alfa Aesar™
J6057422
100g Each for $28.38
 
AAJ6057436 View Documents Alfa Aesar™
J6057436
500g Each for $98.24
 
Description

Description

C.I. 11160
Specifications

Chemical Identifiers

97-56-3
225.295
PFRYFZZSECNQOL-UHFFFAOYSA-N
7340
CC1=CC=CC=C1N=NC2=CC(=C(C=C2)N)C
C14H15N3
MFCD00007733
C.I. 11160; Solvent Yellow 3
2-methyl-4-[(2-methylphenyl)diazenyl]aniline

Specifications

97-56-3
CH3C6H4N=NC6H3(CH3)NH2
6506005
C.I. 11160; Solvent Yellow 3
CC1=CC=CC=C1N=NC2=CC(=C(C=C2)N)C
225.295
225.29
Odorless
Brown-Orange
Fast Garnet GBC base
C14H15N3
MFCD00007733
14,420
PFRYFZZSECNQOL-UHFFFAOYSA-N
2-methyl-4-[(2-methylphenyl)diazenyl]aniline
7340
Powder
101°C to 102°C
100g
Safety and Handling

Safety and Handling

GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : XU8800000

TSCA : TSCA