missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fast Garnet GBC base, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J6057422

 View more versions of this product

Catalog No. AAJ6057422



Description

Description

C.I. 11160

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C.I. 11160
Specifications

Specifications

Fast Garnet GBC base
97-56-3
CH3C6H4N=NC6H3(CH3)NH2
6506005
C.I. 11160; Solvent Yellow 3
CC1=CC=CC=C1N=NC2=CC(=C(C=C2)N)C
225.295
225.29
Brown/Orange
Powder
101°C to 102°C
C14H15N3
MFCD00007733
14,420
PFRYFZZSECNQOL-UHFFFAOYSA-N
2-methyl-4-[(2-methylphenyl)diazenyl]aniline
7340
Odorless
100 g

Chemical Identifiers

97-56-3
225.295
PFRYFZZSECNQOL-UHFFFAOYSA-N
7340
CC1=CC=CC=C1N=NC2=CC(=C(C=C2)N)C
C14H15N3
MFCD00007733
C.I. 11160; Solvent Yellow 3
2-methyl-4-[(2-methylphenyl)diazenyl]aniline
Safety and Handling

Safety and Handling

GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.

P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H350

EINECSNumber : 202-591-2

RTECSNumber : XU8800000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only