missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fast blue B salt, MP Biomedicals™ DFS Item

$167.88 - $167.88

Chemical Identifiers

CAS 14263-94-6
Molecular Formula C14H12Cl4N4O2Zn
Molecular Weight (g/mol) 475.46
InChI Key GPPKNJIWDULNQH-UHFFFAOYSA-J
Synonym AZOIC DIAZO No 48, C.I. 37235
PubChem CID 66685
ChEBI CHEBI:87629
IUPAC Name zinc(2+) 3,3'-dimethoxy-[1,1'-biphenyl]-4,4'-bis(diazonium) tetrachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Zn++].COC1=CC(=CC=C1[N+]#N)C1=CC=C([N+]#N)C(OC)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN15020110
View Documents
Mp Biomedicals
0215020110
10g
Each for $167.88
 
Description

Description

Specifications

Chemical Identifiers

14263-94-6
475.46
AZOIC DIAZO No 48, C.I. 37235
CHEBI:87629
[Cl-].[Cl-].[Cl-].[Cl-].[Zn++].COC1=CC(=CC=C1[N+]#N)C1=CC=C([N+]#N)C(OC)=C1
C14H12Cl4N4O2Zn
GPPKNJIWDULNQH-UHFFFAOYSA-J
66685
zinc(2+) 3,3'-dimethoxy-[1,1'-biphenyl]-4,4'-bis(diazonium) tetrachloride

Specifications

14263-94-6
AZOIC DIAZO No 48, C.I. 37235
[Cl-].[Cl-].[Cl-].[Cl-].[Zn++].COC1=CC(=CC=C1[N+]#N)C1=CC=C([N+]#N)C(OC)=C1
475.46
CHEBI:87629
Powder
10g
C14H12Cl4N4O2Zn
GPPKNJIWDULNQH-UHFFFAOYSA-J
zinc(2+) 3,3'-dimethoxy-[1,1'-biphenyl]-4,4'-bis(diazonium) tetrachloride
66685
475.5
Purple
Fast blue B salt
SDS