missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Fascin Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Fascin Ab-1, Mouse Monoclonal Antibody.

$390.76 - $1,272.78

Specifications

Antigen Fascin Ab-1
Clone FCN01; same as 55K-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MS1112R7 Thermo Scientific™ Lab Vision™
MS1112R7
7mL Each for $889.90
MS1112S Thermo Scientific™ Lab Vision™
MS1112S
1mL Each for $1,272.78
MS1112S0 Thermo Scientific™ Lab Vision™
MS1112S0
100μL Each for $390.76
MS1112S1 Thermo Scientific™ Lab Vision™
MS1112S1
500μL Each for $899.47
Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Fascin Ab-1, Mouse Monoclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Human fascin is a highly conserved actin-bundling protein. Fascin, encoded by the human homolog for sn (hsn) gene, has been localized to microspikes and stress fibers of cultured cells where it is thought to be involved in the formation of microfilament bundles. It is expressed predominantly in dendritic cells. Lymphoid cells, myeloid cells and plasma cells are negative. However, Reed Sternberg cells in Hodgkin's lymphoma are positive for fascin staining. Epstein-Barr virus may induce expression of fascin in B cells.

Host Species: Mouse

Clone: FCN01; same as 55K-2

Isotype: IgG1

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Fascin purified from HeLa cells

Molecular Weight: 55 kDa

Positive Control: Hodgkin's lymphoma or spleen

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Description & Specifications

Specifications

Fascin Ab-1
Immunohistochemistry (Paraffin)
Unlabeled
IVD
Primary
FCN01; same as 55K-2
Monoclonal
Murine
Neuroscience
Fascin purified from HeLa cells