Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Fas Ligand, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Fas Ligand, Rabbit Polyclonal Antibody.

$301.49 - $1,132.04

Specifications

Antigen Fas Ligand
Applications Western Blot
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

RB9029P

 
thermo scientific™ lab vision™
RB9029P
1mL Each for $1,132.04

RB9029P0

 
thermo scientific™ lab vision™
RB9029P0
100μL Each for $301.49

RB9029P1

 
thermo scientific™ lab vision™
RB9029P1
500μL Each for $754.18

RB9029R7

 
thermo scientific™ lab vision™
RB9029R7
7mL Each for $724.16
Description & Specifications

Specifications

Antigen Fas Ligand
Applications Western Blot
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Host Species Rabbit
Regulatory Status RUO
Research Discipline Apoptosis
Type Primary
Immunogen A synthetic peptide derived from N-terminal of human Fas ligand
Isotype IgG

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Fas Ligand, Rabbit Polyclonal Antibody.

Fas ligand is a cell surface molecule belonging to the tumor necrosis factor family,. It binds to its receptor Fas, thus inducing apoptosis. Various cells express Fas, where Fas ligand is expressed predominantly on activated T cells. Fas and Fas ligand are involved in the down-regulation of immune reactions as well as T cell-mediated cytotoxicity.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: N-terminus

Immunogen: A synthetic peptide derived from N-terminal of human Fas ligand

Molecular Weight: 37 kDa

Positive Control: Tonsil

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO