Please login to your online account to display your discounted pricing

Alfa Aesar™ Faropenem sodium salt hydrate


Quantity 50mg

Manufacturer: alfa aesar™  J66803-MF

 View more versions of this product

Catalog No. AAJ66803MF

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Color Off-white to pale brown
Quantity 50mg
Chemical Name or Material Faropenem sodium salt hydrate
CAS 122547-49-3
Physical Form Powder
Formula Weight 307.30g/mol (Anhydrous)
MDL Number MFCD17215925
Merck Index 14,3938
Synonym ALP 201; Farom
Solubility Information Soluble in water
Storage Note 1 -30° to -10°C
RTECS Number XI0198400

Specifications

Chemical Name or Material Faropenem sodium salt hydrate
CAS 122547-49-3
Formula Weight 307.30g/mol (Anhydrous)
MDL Number MFCD17215925
Merck Index 14,3938
Synonym ALP 201; Farom

  • A broad spectrum antibiotic and has shown to be very effective against anaerobic pathogens known to cause oral infections

Accessories

-30° to -10°C