Please login to your online account to display your discounted pricing

Alfa Aesar™ Faropenem sodium salt hydrate

Chemical Identifiers

Chemical Name or Material Faropenem sodium salt hydrate
CAS 122547-49-3
Formula Weight 307.30g/mol (Anhydrous)
MDL Number MFCD17215925
Merck Index 14,3938
Synonym ALP 201; Farom
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66803MF

 
alfa aesar™
J66803-MF
50mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Off-white to pale brown
Quantity 50mg
Chemical Name or Material Faropenem sodium salt hydrate
CAS 122547-49-3
Physical Form Powder
Formula Weight 307.30g/mol (Anhydrous)
MDL Number MFCD17215925
Merck Index 14,3938
Synonym ALP 201; Farom
Solubility Information Soluble in water
Storage Note 1 -30° to -10°C
RTECS Number XI0198400

  • A broad spectrum antibiotic and has shown to be very effective against anaerobic pathogens known to cause oral infections

Safety and Handling

-30° to -10°C