Please login to your online account to display your discounted pricing

Alfa Aesar™ Farnesylthioacetic acid, 98%


Quantity 5mg

Manufacturer: alfa aesar™  J67240LB0

 View more versions of this product

Catalog No. AAJ67240LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 296.47
Chemical Name or Material Farnesylthioacetic acid
Assay 98%
CAS 135784-48-4
Physical Form Oil
MDL Number MFCD00216146
Synonym FTA
Storage Note 1 -30° to -10°C
Color Yellow
Quantity 5mg
Sensitivity Light Sensitive

Specifications

Chemical Name or Material Farnesylthioacetic acid
CAS 135784-48-4
Formula Weight 296.47
MDL Number MFCD00216146
Synonym FTA
Accessories

-30° to -10°C