Please login to your online account to display your discounted pricing

Alfa Aesar™ Farnesylthioacetic acid, 98%

Chemical Identifiers

Chemical Name or Material Farnesylthioacetic acid
CAS 135784-48-4
Formula Weight 296.47
MDL Number MFCD00216146
Synonym FTA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67240LB0

 
alfa aesar™
J67240LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 296.47
CAS 135784-48-4
Chemical Name or Material Farnesylthioacetic acid
Assay 98%
MDL Number MFCD00216146
Synonym FTA
Physical Form Oil
Quantity 5mg
Color Yellow
Storage Note 1 -30° to -10°C
Sensitivity Light Sensitive
Safety and Handling

-30° to -10°C