missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Falcon™ Tissue Culture Treated Flasks

Manufactured from optically clear virgin polystyrene for easy viewing of sample or cells. Corning™ Falcon™ Tissue Culture Treated Flasks features vacuum-gas plasma tissue culture treatment to ensure consistent growth surface.

Manufacturer:  Corning Life Sciences C353135

 View more versions of this product

Catalog No. 13-680-59



Description

Description

 

  • Nonpyrogenic; Sterile
  • Manufactured under a registered ISO 9002 quality system
  • 100% leak testing of injection-molded flasks
  • Volumetric graduation accuracy (±5% of volume) Stringent requirements for flatness of optical growth surface
  • Canted-neck 150cm2 flasks which are low-profile for stacking and efficient use in incubator; include locking caps and graduations
  • Straight-neck 175cm2 flasks have frosted writing area
  • 225cm2 flasks with locking caps, frosted writing area, vertical graduations, 100mL maximum horizontal working volume line and a skirted, canted neck for extra stability
  • 300cm2 flasks, manufactured in an ISO Class 8 cleanroom with blow-molded design, printed graduations, straight neck, and white marking area
Cap Types
  • Two-position plug-seal caps for use in open or closed incubation Vented caps incorporate a 0.2μm membrane for consistent gas exchange and reduced contamination Rigid phenolic (lined) caps twist on and provide the most reliable liquid/gastight sealing
  • Patented locking incubation position, found on the 150cm2 and 225cm2 flasks, prevents caps from falling off or closing while in the open position

 

Specifications

Specifications

8.45 oz.
Plug-seal
250mL
For enhanced cell attachment and growth
Rectangular
Tissue culture treated
Polyethylene
75cm2
PS (Polystyrene)
Canted
Sterile
Documents